Learn More
Restek™ Flip Seal™ Dual Vespel™ Ring Inlet Seals
Reversible, two-sided design allows significantly more analyses than other seals
Brand: Restek™ 23412
This item is not returnable.
View return policy
Description
- Highly inert gold or Siltek™deactivated seals reduce breakdown and adsorption of active compounds, maximizing component transfer to GC column
- Vespel™ ring embedded in top and bottom surface eliminates need for washer
- Very little torque required to make seal, reduces operator variability
Specifications
Siltek-Deactivated | |
1.2 mm | |
Ded Design Allows Significantly More Analyses Than Other Seals | |
2/Pk. |