Learn More
Restek™ Flip Seal™ Dual Vespel™ Ring Inlet Seals
Reversible, two-sided design allows significantly more analyses than other seals
£76.23 - £375.10
Specifications
Diameter (Metric) Inner | 1.2 mm |
---|---|
For Use With (Application) | Ded Design Allows Significantly More Analyses Than Other Seals |
Product Code | Brand | Diameter (Metric) Inner | Type | Packaging Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Type | Packaging Quantity | Price | Quantity & Availability | |||
13139912
|
Restek™
23414 |
1.2 mm | Siltek-Deactivated | 10/Pk. |
£375.10
Pack of 10 |
Estimated Shipment: 14-02-2025
Log in to see stock. |
|||
13119912
|
Restek™
23412 |
1.2 mm | Siltek-Deactivated | 2/Pk. |
£76.23
Pack of 2 |
Estimated Shipment: 14-02-2025
Log in to see stock. |
|||
13109912
|
Restek™
23411 |
1.2 mm | Gold-Plated | 2/Pk. |
£82.28
Pack of 2 |
Estimated Shipment: 14-02-2025
Log in to see stock. |
|||
13129912
|
Restek™
23413 |
1.2 mm | Gold-Plated | 10/Pk. |
£370.26
Pack of 10 |
Estimated Shipment: 14-02-2025
Log in to see stock. |
|||
Description
- Highly inert gold or Siltek™deactivated seals reduce breakdown and adsorption of active compounds, maximizing component transfer to GC column
- Vespel™ ring embedded in top and bottom surface eliminates need for washer
- Very little torque required to make seal, reduces operator variability
Specifications
1.2 mm | |
Ded Design Allows Significantly More Analyses Than Other Seals |