Learn More
Cole-Parmer™ Stuart™ Florentine Flask, Plastic-coated For Stuart™ Rotary Evaporators
Plastic coating prevents shattering. Used with Stuart™ Rotary Evaporators.
Brand:  Cole-Parmer™ 10096-59
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12417742
  • £362.90 / Each
Estimated Shipment: 05-08-2024
to see stock.
Add to basket
Description
Description
  • For use with Stuart™ rotary evaporators
  • Plastic coated glass
  • 29/32 socket
Specifications
Specifications
Evaporating flask
Plastic-coated Glass
Flat
2 L
29/32