Learn More
Florisil™, for pesticide residue analysis, 60-100 mesh
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS | 1343-88-0 |
Quantity | 500 g |
Molecular Formula | MgO3Si |
InChI Key | ZADYMNAVLSWLEQ-UHFFFAOYSA-N |
Solubility | Insoluble in water |
Molecular Weight (g/mol) | 100.39 |
Formula Weight | 100.39 |
Grade | Pesticide |
Chemical Name or Material | Florisil™ |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0464576 | Certificate of Analysis | 23/07/2024 | 457725000 |
A0463239 | Certificate of Analysis | 03/06/2024 | 457725000 |
A0459240 | Certificate of Analysis | 10/01/2024 | 457725000 |
A0459052 | Certificate of Analysis | 04/01/2024 | 457725000 |
A0450702 | Certificate of Analysis | 02/05/2023 | 457725000 |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
1Hours
16Minutes
58Seconds
29
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.