Learn More
Invitrogen™ eBioscience™ Flow Cytometry Staining Buffer
A buffered saline solution containing fetal bovine serum and sodium azide (0.09%) as a preservative. Designed for use in immunofluorescent staining protocols of cells in suspension.
Brand: Invitrogen™ 00-4222-57
43.05 GBP valid until 2025-03-16
Use promo code "24070" to get your promotional price.
Alert:
Contains Sodium azide
Description
- Designed for use in immunofluorescent staining protocols of cells in suspension
- A buffered saline solution containing fetal bovine serum and sodium azide (0.09%) as a preservative
- This buffer can be used for antibody and cell dilution steps, as well as all the wash steps required for the surface staining and flow cytometric analysis
- Contains animal serum proteins to help minimizing non-specific binding of antibodies

Specifications
200 mL | |
eBioscience™ | |
Staining Buffer |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2510322 | Certificate of Analysis | 29/01/2025 | 00-4222-57 |
2481442 | Certificate of Analysis | 29/01/2025 | 00-4222-57 |
2548851 | Certificate of Analysis | 29/01/2025 | 00-4222-57 |
2592922 | Certificate of Analysis | 29/01/2025 | 00-4222-57 |
2822760 | Certificate of Analysis | 26/01/2025 | 00-4222-57 |
Special Offers
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24154Expires: 2025-02-28
Days
8Hours
8Minutes
23Seconds
47
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.