Learn More
Fluorene, 98+%, Thermo Scientific Chemicals
£18.80 - £144.00
Chemical Identifiers
CAS | 86-73-7 |
---|---|
Molecular Formula | C13H10 |
Molecular Weight (g/mol) | 166.22 |
MDL Number | MFCD00001111 |
InChI Key | NIHNNTQXNPWCJQ-UHFFFAOYSA-N |
Synonym | fluorene, diphenylenemethane, o-biphenylenemethane, 2,3-benzindene, 2,2'-methylenebiphenyl, o-biphenylmethane, methane, diphenylene, fluoren, fluorenyl radical, flourene |
PubChem CID | 6853 |
ChEBI | CHEBI:28266 |
IUPAC Name | 9H-fluorene |
SMILES | C1C2=CC=CC=C2C2=CC=CC=C12 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10573321
|
Thermo Scientific Acros
156130250 |
25 g | Plastic bottle |
£18.80
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10513901
|
Thermo Scientific Acros
156131000 |
100 g | Plastic bottle |
£43.80
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10437750
|
Thermo Scientific Acros
156135000 |
500 g | Plastic bottle |
£144.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
86-73-7 | |
166.22 | |
NIHNNTQXNPWCJQ-UHFFFAOYSA-N | |
6853 | |
9H-fluorene |
C13H10 | |
MFCD00001111 | |
fluorene, diphenylenemethane, o-biphenylenemethane, 2,3-benzindene, 2,2'-methylenebiphenyl, o-biphenylmethane, methane, diphenylene, fluoren, fluorenyl radical, flourene | |
CHEBI:28266 | |
C1C2=CC=CC=C2C2=CC=CC=C12 |
Specifications
86-73-7 | |
Brown to White | |
298.0°C | |
Authentic | |
C13H10 | |
05, 625 | |
1.2 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,benzene and carbon disu,freely soluble in glacial acetic acid | |
C1C2=CC=CC=C2C2=CC=CC=C12 | |
166.22 | |
CHEBI:28266 | |
98+% | |
Fluorene, 98% |
112.0°C to 116.0°C | |
1.2000g/mL | |
151°C | |
98% min. (GC) | |
MFCD00001111 | |
15, 4187 | |
fluorene, diphenylenemethane, o-biphenylenemethane, 2,3-benzindene, 2,2'-methylenebiphenyl, o-biphenylmethane, methane, diphenylene, fluoren, fluorenyl radical, flourene | |
NIHNNTQXNPWCJQ-UHFFFAOYSA-N | |
9H-fluorene | |
6853 | |
166.22 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 201-695-5
missing translation for 'rtecsNumber' : LL5670000
missing translation for 'tsca' : TSCA
- Fluorene
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
Hazard Statement
- H410-Very toxic to aquatic life with long lasting effects.
- H400-Very toxic to aquatic life.
Precautionary Statement
- P273-Avoid release to the environment.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.