missing translation for 'onlineSavingsMsg'
Learn More

Fluorescamine, pure, Thermo Scientific Chemicals

£32.20 - £1047.00

Chemical Identifiers

CAS 38183-12-9
Molecular Formula C17H10O4
Molecular Weight (g/mol) 278.26
MDL Number MFCD00005928
InChI Key ZFKJVJIDPQDDFY-UHFFFAOYNA-N
PubChem CID 37927
IUPAC Name 4'-phenylspiro[2-benzofuran-3,2'-furan]-1,3'-dione
SMILES O=C1OC2(OC=C(C2=O)C2=CC=CC=C2)C2=CC=CC=C12
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10144450
View Documents Promotion Details
Thermo Scientific Acros
191670250
25 mg Glass bottle
£32.20
25mg
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
10153730
View Documents Promotion Details
Thermo Scientific Acros
191671000
100 mg Glass bottle
£78.80
100mg
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
10551831
View Documents Promotion Details
Thermo Scientific Acros
191675000
500 mg Glass bottle
£316.00
500mg
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
11946201
View Documents Promotion Details
Thermo Scientific Acros
191670025
2.5 g Glass bottle
£1047.00
2.50g
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

38183-12-9
278.26
ZFKJVJIDPQDDFY-UHFFFAOYNA-N
4'-phenylspiro[2-benzofuran-3,2'-furan]-1,3'-dione
C17H10O4
MFCD00005928
37927
O=C1OC2(OC=C(C2=O)C2=CC=CC=C2)C2=CC=CC=C12

Specifications

155.0°C to 157.0°C
C17H10O4
Solubility in water: .. Other solubilities: soluble in diethyl ether
Authentic
4'-phenylspiro[2-benzofuran-3,2'-furan]-1,3'-dione
37927
Pure
Yellow
Fluorescamine
38183-12-9
MFCD00005928
ZFKJVJIDPQDDFY-UHFFFAOYNA-N
O=C1OC2(OC=C(C2=O)C2=CC=CC=C2)C2=CC=CC=C12
278.26
278.26
900 (235nm, ether) min.
Powder
Videos
Safety and Handling

Safety and Handling

Warning

Product Identifier
  • Fluorescamine

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only