missing translation for 'onlineSavingsMsg'
Learn More

Falcon™ FluoroBlok™ 24 Well Permeable Supports and Companion Plates
GREENER_CHOICE

Block light transmission, simplifying detection of migrating fluorescently labeled cells. Falcon™ FlouroBlok™ 24 Well Plate Permeable Supports are designed with a unique light-tight PET membrane. Must be used with Falcon™ 24 Well Companion Plate.

Brand:  Falcon™ 351152

 View more versions of this product

Product Code. 10365961

  • £425.91 / Pack of 48
Estimated Shipment: 24-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Falcon Cell Culture Insert Companion Plates have been specially designed for use with Falcon or Corning™ BioCoatTM Cell Culture Inserts so evaporation and contamination due to improper lid fit is eliminated.

In the Feeding Position, pipet access is improved for fluid handling on the basolateral side. Reagents can be added quickly and consistently for timed experiments. Aspiration of media from the well is easier, reducing the risk of contamination.

In the Incubation Position, Falcon Cell Culture Inserts remain locked in position in their Falcon Companion Plate wells. Media will not wick up between the insert and well wall. The unique Falcon low-evaporation lid provides a tortuous air passage system that reduces evaporation and contamination.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cell Culture Insert, 8 μm Pore Size
PET
Cell Culture
24
48/Cs.
8 μm
0.65 cm
0.3 cm2
Sterile
Multi-well Insert System
Falcon™ 24 Well TC-Treated Cell Polystyrene Permeable Support Companion Plate
8/Pk.
Polyethylene Terephthalate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.