Learn More
Falcon™ FluoroBlok™ 24 Well Permeable Supports and Companion Plates
Block light transmission, simplifying detection of migrating fluorescently labeled cells. Falcon™ FlouroBlok™ 24 Well Plate Permeable Supports are designed with a unique light-tight PET membrane. Must be used with Falcon™ 24 Well Companion Plate.
£175.00 - £450.00
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | ||||
10375911
|
Falcon™
351151 |
Cell Culture Insert, 3 μm Pore Size | Falcon™ 24 Well TC-Treated Cell Polystyrene Permeable Support Companion Plate |
£450.00
Pack of 48 |
In Stock
Log in to see stock. |
||||
10365961
|
Falcon™
351152 |
Cell Culture Insert, 8 μm Pore Size | Falcon™ 24 Well TC-Treated Cell Polystyrene Permeable Support Companion Plate |
£425.91
Pack of 48 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
10132371
|
Falcon™
351150 |
Cell Culture Insert; 1 μm pore size | Falcon™ 24 Well TC-Treated Cell Polystyrene Permeable Support Companion Plate | N/A | |||||
10459412
|
Falcon™
353504 |
Insert Companion Plate | Falcon™ or Corning™ BioCoat™ Cell culture Inserts; may be used with or without cell culture inserts |
£175.00
Pack of 50 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||
Description
Falcon Cell Culture Insert Companion Plates have been specially designed for use with Falcon or Corning™ BioCoatTM Cell Culture Inserts so evaporation and contamination due to improper lid fit is eliminated.
In the Feeding Position, pipet access is improved for fluid handling on the basolateral side. Reagents can be added quickly and consistently for timed experiments. Aspiration of media from the well is easier, reducing the risk of contamination.
In the Incubation Position, Falcon Cell Culture Inserts remain locked in position in their Falcon Companion Plate wells. Media will not wick up between the insert and well wall. The unique Falcon low-evaporation lid provides a tortuous air passage system that reduces evaporation and contamination.
Specifications
Sterile | |
24 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.