Learn More
Invitrogen™ FluoSpheres™ Carboxylate-Modified Microspheres
Achieve the brightest fluorescence with Carboxylate-Modified FluoSphere Microspheres, available in different colors and particle sizes.
£409.00 - £656.00
Specifications
For Use With (Application) | Fluorescence Microscopy |
---|---|
Material | Polystyrene |
Product Type | Carboxylate-Modified Microsphere |
Product Line | FLUOSPHERES™ |
Surface Modification | Carboxylate |
Product Code | Brand | Diameter (Metric) | Color | Excitation/Emission | Quantity | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Color | Excitation/Emission | Quantity | Price | Quantity & Availability | ||
10328192
|
Invitrogen™
F8803 |
0.1 μm | Yellow-Green | 505/515 nm | 10 mL |
£460.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10696523
|
Invitrogen™
F8816 |
1.0 μm | Crimson | 625/645 nm | 2 mL |
£451.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10023162
|
Invitrogen™
F8823 |
1.0 μm | Yellow-Green | 505/515 nm | 10 mL |
£463.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10484962
|
Invitrogen™
F8801 |
0.1 μm | Red | 580/605 nm | 10 mL |
£469.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10513463
|
Invitrogen™
F8811 |
0.2 μm | Yellow-Green | 505/515 nm | 10 mL |
£456.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10033062
|
Invitrogen™
F8807 |
0.2 μm | Dark Red | 660/680 nm | 2 mL |
£458.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10573663
|
Invitrogen™
F10720 |
0.04 μm | Yellow-Green, Orange, Red, Dark Red | 505/515, 540/560, 580/605, 660/680 nm | 1 mL/each |
£656.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11514796
|
Invitrogen™
F20881 |
0.2 μm | Orange | 365/610 nm | 2 mL |
£415.00
2mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10751154
|
Invitrogen™
F8783 |
0.02 μm | Dark Red | 660/680 nm | 2 mL |
£427.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10398352
|
Invitrogen™
F8786 |
0.02 μm | Red | 580/605 nm | 10 mL |
£424.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10338652
|
Invitrogen™
F8795 |
0.04 μm | Yellow-Green | 505/515 nm | 1 mL |
£442.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10462472
|
Invitrogen™
F8813 |
0.5 μm | Yellow-Green | 505/515 nm | 10 mL |
£450.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10318532
|
Invitrogen™
F8820 |
1.0 μm | Orange | 540/560 nm | 10 mL |
£421.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10207002
|
Invitrogen™
F8825 |
2.0 μm | Nile Red | 535/575 nm | 2 mL |
£409.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10768424
|
Invitrogen™
F8827 |
2.0 μm | Yellow-Green | 505/515 nm | 2 mL |
£455.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10043302
|
Invitrogen™
F8781 |
0.02 μm | Blue | 365/415 nm | 10 mL |
£419.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10684243
|
Invitrogen™
F8782 |
0.02 μm | Crimson | 625/645 nm | 2 mL |
£414.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10761154
|
Invitrogen™
F8784 |
0.02 μm | Nile Red | 535/575 nm | 10 mL |
£451.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10412892
|
Invitrogen™
F8787 |
0.02 μm | Yellow-Green | 505/515 nm | 10 mL |
£449.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10093262
|
Invitrogen™
F8789 |
0.04 μm | Dark Red | 660/680 nm | 1 mL |
£463.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10378932
|
Invitrogen™
F8792 |
0.04 μm | Orange | 540/560 nm | 1 mL |
£446.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10606553
|
Invitrogen™
F8793 |
0.04 μm | Red | 580/605 nm | 1 mL |
£450.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10144722
|
Invitrogen™
F8794 |
0.04 μm | Red-Orange | 565/580 nm | 1 mL |
£409.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10003572
|
Invitrogen™
F8797 |
0.1 μm | Blue | 350/440 nm | 10 mL |
£421.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10154722
|
Invitrogen™
F8799 |
0.1 μm | Infrared | 715/755 nm | 1 mL |
£411.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10043142
|
Invitrogen™
F8800 |
0.1 μm | Orange | 540/560 nm | 10 mL |
£417.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10296612
|
Invitrogen™
F8805 |
0.2 μm | Blue | 365/415 nm | 10 mL |
£441.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10463622
|
Invitrogen™
F8806 |
0.2 μm | Crimson | 625/645 nm | 2 mL |
£444.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10328482
|
Invitrogen™
F8809 |
0.2 μm | Orange | 540/560 nm | 10 mL |
£448.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10544803
|
Invitrogen™
F8810 |
0.2 μm | Red | 580/605 nm | 10 mL |
£460.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10738044
|
Invitrogen™
F8812 |
0.5 μm | Red | 580/605 nm | 10 mL |
£451.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10398142
|
Invitrogen™
F8814 |
1.0 μm | Blue | 365/415 nm | 10 mL |
£424.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10358482
|
Invitrogen™
F8815 |
1.0 μm | Blue | 350/440 nm | 10 mL |
£414.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10217142
|
Invitrogen™
F8819 |
1.0 μm | Nile Red | 535/575 nm | 10 mL |
£417.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10564423
|
Invitrogen™
F8821 |
1.0 μm | Red | 580/605 nm | 10 mL |
£423.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10758244
|
Invitrogen™
F8824 |
2.0 μm | Blue | 365/415 nm | 2 mL |
£409.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10318632
|
Invitrogen™
F8826 |
2.0 μm | Red | 580/605 nm | 2 mL |
£426.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Easily perform flow cytometry, microscopy, HTS, HCS, immunoassay, and other laboratory applications using our extensive selection of FluoSpheres Carboxylate-Modified Microspheres. FluoSphere beads can be used in passive adsorption or active, covalent coupling of proteins, nucleic acids, and biomolecules for particle capture applications. FluoSphere microspheres are loaded with proprietary fluorescent dyes, making them the brightest microspheres available.
Visualize the brightest fluorescence for laboratoy applications including fluorescence microscopy, flow cytometry, HTS, HCS, and cell tracing with our Carboxylate-Modified FluoSphere Microspheres, which are manufactured from polystyrene microspheres and loaded with different proprietary dyes. Using specialized staining methods enables all of the fluorescent dye molecules to be contained inside each polystyrene microsphere instead of on the bead's surface. This protective environment within the bead shields the dye from detrimental environmental effects, such as photobleaching. Our carboxylate-modified microspheres are coated with a hydrophilic polymer containing multiple carboxylic acids for covalent attachment of ligands. A range of particle sizes is available for different research uses and experiments.Specifications
Fluorescence Microscopy | |
Carboxylate-Modified Microsphere | |
Carboxylate |
Polystyrene | |
FLUOSPHERES™ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.