missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One FLUOTRAC™ 200 96-well Non-Treated Microplate
For use in fluorescence immunoassay. Greiner Bio-One FLUOTRAC™ 200 96-Well Non-Treated Microplate is suitable for non polar proteins and peptides.
Brand: Greiner Bio-One 675075
This item is not returnable.
View return policy
Description
- Free of detectable DNase, RNase, human DNA Non-pyrogenic
- Manufactured under DIN ISO 9001 guidelines
- Can be traced back to production through defined LOT number system
- Reduction of sample volume by up to 50 %
- Sta
Service Plan
No
Services
No
Specifications
No | |
Half-Area | |
8 x 12 | |
Medium Binding | |
Polystyrene | |
Non-sterile | |
199 μL | |
40 | |
40/Cs. | |
White |
Medium Binding | |
Without Lid | |
Not Autoclavable | |
White | |
96 | |
Non-treated | |
Round, Flat Bottom (F-well) | |
10 | |
Yes |