missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One FLUOTRAC™ 200 96-well Non-Treated Microplate
For use in fluorescence immunoassay. Greiner Bio-One FLUOTRAC™ 200 96-Well Non-Treated Microplate is suitable for non polar proteins and peptides.
£220.00 - £232.00
Specifications
Format | Half-Area |
---|---|
Array | 8 x 12 |
Material | Polystyrene |
No. of Wells | 96 |
Surface Treatment | Non-treated |
Product Code | Brand | Color | Binding Property | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Binding Property | Price | Quantity & Availability | ||||
10105423
|
Greiner Bio-One
675075 |
White | Medium Binding |
£220.00
Pack of 40 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10023973
|
Greiner Bio-One
675074 |
White | High Binding |
£232.00
Pack of 40 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10701634
|
Greiner Bio-One
675077 |
Black | High Binding |
£220.00
Pack of 40 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Free of detectable DNase, RNase, human DNA Non-pyrogenic
- Manufactured under DIN ISO 9001 guidelines
- Can be traced back to production through defined LOT number system
- Reduction of sample volume by up to 50 %
- Sta
Service Plan
No
Warranty
No
Specifications
Half-Area | |
Polystyrene | |
Non-treated | |
40/Cs. |
8 x 12 | |
96 | |
40 |