missing translation for 'onlineSavingsMsg'
Learn More

Keystone Cleanroom Products Cleanroom Face Masks, Gamma Irradiated
GREENER_CHOICEBIOPHARMA

Gamma irradiated cleanroom facemasks eliminate goggles from fogging and mask slippage. Available with ties, headband, or earloop fastening.

Brand:  Keystone Cleanroom Products S-1461DEL2

 View more versions of this product

Product Code. 17725299

  • £2375.00 / Pack of 500
Estimated Shipment: 10-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Sterile cleanroom face masks
  • Certificates of Irradiation and Conformance with each shipment
  • Fastening Types: Earloops, Ties, or Headband
Specifications

Specifications

Cleanroom Face Mask
Headband
Large
7 x 4 in.
Class 100 (ISO 5)
Yes
18 cm
500/Case
10 cm
Blue
SMS
Sterile
Pleated
Single-use
7 in.
S-1461
4 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers