missing translation for 'onlineSavingsMsg'
Learn More

Keystone Cleanroom Products Cleanroom Face Masks, Gamma Irradiated with Tyvek™ Side Extension
GREENER_CHOICEBIOPHARMA

Sterile cleanroom facemasks feature an tyvek side extension for additional protection. Available with ties or double elastic headband fastening.

Brand:  Keystone Cleanroom Products S-1461XLDEL2AD

 View more versions of this product

Product Code. 17765299

  • £3430.00 / Pack of 500
Estimated Shipment: 19-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Sterile cleanroom face masks
  • Certificates of Irradiation and Conformance with each shipment
  • Tyvek™ side extensions allow mask expansion to increase coverage and prevent unwanted facial skin exposure
Specifications

Specifications

Cleanroom Face Mask
Headband
X-Large
12.25 x 4 in.
Class 100 (ISO 5)
Side Extension
12.25 in.
S-1461
10 cm
Green
SMS
Sterile
Pleated
Single-use
Yes
31 cm
4 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers