Learn More
BRAND™ Forceps
PTFE forceps
Brand:  BRAND™ 113130
 View more versions of this product
Product Code. 10746861
  • £48.75 / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Rectangular ends
  • Outstanding chemical and thermal resistance
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Rectangular
    PTFE
    150 mm
    1