man
Test
Search
Order Status
Quick Order
Sign In
chevron-down
chevron-down
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Surgical Tools
Forceps and Tweezers
BRAND™ Forceps
Print
BRAND™ Forceps
PTFE forceps
£48.25 - £73.95
See promotional offers below
Specifications
Tip Style
Rectangular
Material
PTFE
Quantity
1
View More Specs
Products
2
Description
Specifications
Videos
SDS
Documents
Product Certifications
Products
2
Product Code
Brand
Length (Metric)
Price
Quantity & Availability
Product Code
Brand
Length (Metric)
Price
Quantity & Availability
10263111
View Documents
Promotion Details
missing translation for 'viewProductCertificates'
BRAND™
113135
200 mm
£73.95
Each
View alternative products
Save Up to
»
Estimated Shipment: 10-09-2024
Log in
to see stock.
Add to basket
10746861
View Documents
Promotion Details
missing translation for 'viewProductCertificates'
BRAND™
113130
150 mm
£48.25
Each
View alternative products
Save Up to
»
In Stock
Log in
to see stock.
Add to basket
Description
Description
Rectangular ends
Outstanding chemical and thermal resistance
Specifications
Specifications
Tip Style
Rectangular
Material
PTFE
Quantity
1
Videos
SDS
Documents
Documents
Product Certifications
Back to Top