Learn More
Thermo Scientific™ Forma™ Sample Probe Kits
Measure sample temperature or top chamber temperature in upright freezers with Thermo Scientific™ Forma™ Sample Probe Kits.
Brand: Thermo Scientific™ 195892
Code : NEW
Additional Details : Weight : 0.80000kg
Description
Measure sample temperature or top chamber temperature in upright freezers with Thermo Scientific™ Forma™ Sample Probe Kits. Use probe kit with Forma 8600 Series Upright Freezers.
- Useful for measuring sample temperature or top chamber temperature
- Kits consist of probe and probe kit
Specifications
Customer installed | |
Sample Probe | |
Upright |