Learn More
Thermo Scientific™ Forma™ Sample Probe Kits
Measure sample temperature or top chamber temperature in upright freezers with Thermo Scientific™ Forma™ Sample Probe Kits.
£271.05 - £273.64
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11766649
|
Thermo Scientific™
195892 |
Customer installed |
£271.05
Each |
Estimated Shipment: 07-10-2024
Log in to see stock. |
|||||
11808003
|
Thermo Scientific™
195893 |
Factory installed |
£273.64
Each |
Estimated Shipment: 07-10-2024
Log in to see stock. |
|||||
Description
Measure sample temperature or top chamber temperature in upright freezers with Thermo Scientific™ Forma™ Sample Probe Kits. Use probe kit with Forma 8600 Series Upright Freezers.
- Useful for measuring sample temperature or top chamber temperature
- Kits consist of probe and probe kit
Specifications
Sample Probe | |
Upright |