missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ Formazin Turbidity Standard, 4000 NTU

Turbidity standard can be diluted to be used with any of Hach's turbidimeters.

Brand:  Hach Lange™ 246149

Product Code. 10045104

  • £212.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Turbidity standard can be diluted to be used with any of Hach's turbidimeters.

    Specifications

  • Concentration: 4000 NTU
  • Parameter: Turbidity
  • Quantity: 500 mL 500 mL
  • Range: N/A
  • Shelf Life: 12 months
  • Storage conditions: Store in a cool, dark place, away from direct sunlight
Specifications

Specifications

Turbidity Standard
Hach Turbidimeters
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hach Lange™ Formazin Turbidity Standard, 4000 NTU >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.