missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
CAS | 64-18-6 |
---|---|
Molecular Formula | CH2O2 |
Molecular Weight (g/mol) | 46.025 |
InChI Key | BDAGIHXWWSANSR-UHFFFAOYSA-N |
Synonym | methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglattShow More |
PubChem CID | 284 |
ChEBI | CHEBI:30751 |
IUPAC Name | formic acid |
SMILES | C(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10705031
|
Thermo Scientific Acros
147930250 |
25 mL | Plastic bottle |
£20.30
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10285711
|
Thermo Scientific Acros
147932500 |
250 mL | Plastic bottle |
£21.84
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10071620
|
Thermo Scientific Acros
147930010 |
1 L | Plastic bottle |
£34.74
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10796341
|
Thermo Scientific Acros
147930025 |
2.5 L | Plastic bottle |
£72.69
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11984881
|
Thermo Scientific Acros
147930100 |
10 L | Plastic drum |
£227.13
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
64-18-6 | |
46.025 | |
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglattShow More | |
CHEBI:30751 | |
C(=O)O |
CH2O2 | |
BDAGIHXWWSANSR-UHFFFAOYSA-N | |
284 | |
formic acid |
Specifications
64-18-6 | |
CH2O2 | |
02, 8 | |
15, 4269 | |
BDAGIHXWWSANSR-UHFFFAOYSA-N | |
formic acid | |
284 | |
46.02 | |
Pure | |
Authentic | |
1.3701 to 1.3721 (20°C, 589nm) | |
Formic acid |
98+% | |
HCO2H | |
01,404; 02,202; 03,147; 04,239; 05,316; 07,160; 08,232; 09,226; 11,243; 13,107; 16,177 | |
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt | |
C(=O)O | |
46.025 | |
CHEBI:30751 | |
≥98.0% | |
1.2200g/mL | |
Liquid | |
44mbar at 20°C |
Safety and Handling
Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Formic acid
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- H302-Harmful if swallowed.
- H226-Flammable liquid and vapour.
- EUH071-Corrosive to the respiratory tract.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Formic acid, 98+%, pure, Thermo Scientific Chemicals