missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente™ Four-Neck Round-Bottom Flasks

Reaction flasks with center neck and with 3 parallel side necks

Brand:  Lenz Laborglasintrumente™ 03345473

Code : A0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11836154

  • £164.30 / Each
Estimated Shipment: 02-08-2024
to see stock.
Add to basket

Description

Description

  • Acc. to DIN 12392 and in addition to DIN standard
  • With 3 parallel side necks
  • With conical ground joints
  • With inscription label
  • Material: DURAN™
Specifications

Specifications

Center neck - NS 45/40, Side necks - NS 29/32
Borosilicate Glass (3.3)
Reaction flasks
2000 mL
1
166 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers