missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12764-1-AP-150UL
This item is not returnable.
View return policy
Description
FOXG1B belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of FOXG1B has not yet been determined; however, it may play a role in the development of the brain and telencephalon. This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of the brain and telencephalon. This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of the brain and telencephalon.Specifications
FOXG1 | |
Polyclonal | |
Unconjugated | |
FOXG1 | |
BF 1, BF 2, BF1, BF2, Brain factor 1, Brain factor 2, FHKL3, FKH2, FKHL1, FKHL2, FKHL3, FKHL4, forkhead box G1, Forkhead box protein G1, Forkhead box protein G1A, Forkhead box protein G1B, Forkhead box protein G1C, Forkhead related protein FKHL1, Forkhead related protein FKHL2, Forkhead related protein FKHL3, FOXG1, FOXG1A, FOXG1B, FOXG1C, HBF 1, HBF 2, HBF 3, HBF G2, HBF2, HFK1, HFK2, HFK3, KHL2, QIN | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15228, 2290, 24370 | |
-20°C | |
Liquid |
Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P55316, Q00939, Q60987 | |
FOXG1 | |
FOXG1 Fusion Protein Ag3426 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FOXG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction