missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | FOXG1 |
---|---|
Concentration | 0.25 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16810664
|
Proteintech
12764-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16800664
|
Proteintech
12764-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FOXG1B belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of FOXG1B has not yet been determined; however, it may play a role in the development of the brain and telencephalon. This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of the brain and telencephalon. This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of the brain and telencephalon.Specifications
FOXG1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
P55316, Q00939, Q60987 | |
15228, 2290, 24370 | |
FOXG1 Fusion Protein Ag3426 | |
Primary | |
-20°C | |
FOXG1 |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
BF 1, BF 2, BF1, BF2, Brain factor 1, Brain factor 2, FHKL3, FKH2, FKHL1, FKHL2, FKHL3, FKHL4, forkhead box G1, Forkhead box protein G1, Forkhead box protein G1A, Forkhead box protein G1B, Forkhead box protein G1C, Forkhead related protein FKHL1, Forkhead related protein FKHL2, Forkhead related protein FKHL3, FOXG1, FOXG1A, FOXG1B, FOXG1C, HBF 1, HBF 2, HBF 3, HBF G2, HBF2, HFK1, HFK2, HFK3, KHL2, QIN | |
FOXG1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |