missing translation for 'onlineSavingsMsg'
Learn More

Human FRAP1 (mTOR), GST Tag Recombinant Protein

Human FRAP1 (mTOR), GST Tag Recombinant Protein

Brand:  Invitrogen PV4753

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10012913

  • £1044.00 / Each
Estimated Shipment: 12-08-2024
to see stock.

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

FRAP1 (mTOR) is a serine⁄threonine kinase that plays a critical role in cellular growth and proliferation. Perturbations in the mTOR⁄PI3-kinase⁄AKT pathway are associated with numerous forms of cancer. FRAP1 is also the target of rapamycin and its analogues, which are currently used as immunosuppressants and cancer therapeutics.
Specifications

Specifications

NP_004949.1
163.9 kDa
FRAP1 (mTOR)
aa 1360-2549
Recombinant
Storage Buffer: 50 mM Tris, pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.1% Polysorbate 20, 2 mM DTT, 50% Glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Ser⁄Thr Kinases: Atypical
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™
FK506 binding protein 12-rapamycin associated protein 1
Kinases & Inhibitors
Liquid
Signal Transduction, Drug Development, Oncology
Kinase Assay
10μg
PIKK Kinase Family
Human
GST Tag
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic
Dry Ice
mTOR, FRAP1
Baculovirus
Recombinant, Catalytic
Serine/Threonine Kinases
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.