missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FRAP1 (mTOR), GST Tag Recombinant Protein
Human FRAP1 (mTOR), GST Tag Recombinant Protein
£1044.00 - £3255.00
Specifications
Accession Number | NP_004949.1 |
---|---|
Gene Alias | FRAP1 (mTOR) |
Species | Human |
Protein Tag | GST Tag |
Expression System | Baculovirus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11380862
|
Invitrogen
PR8683B |
100μg |
£3255.00
Each |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10012913
|
Invitrogen
PV4753 |
10μg |
£1044.00
Each |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
FRAP1 (mTOR) is a serine⁄threonine kinase that plays a critical role in cellular growth and proliferation. Perturbations in the mTOR⁄PI3-kinase⁄AKT pathway are associated with numerous forms of cancer. FRAP1 is also the target of rapamycin and its analogues, which are currently used as immunosuppressants and cancer therapeutics.Specifications
NP_004949.1 | |
Human | |
Baculovirus |
FRAP1 (mTOR) | |
GST Tag |
For Research Use Only. Not for use in diagnostic procedures.