missing translation for 'onlineSavingsMsg'
Learn More

Human FRAP1 (mTOR), GST Tag Recombinant Protein

Human FRAP1 (mTOR), GST Tag Recombinant Protein

Brand:  Invitrogen PR8683B

Code : NEW

Additional Details : Weight : 0.00900kg

 View more versions of this product

Product Code. 11380862

  • £3255.00 / Each
Estimated Shipment: 12-08-2024
to see stock.

Description

Description

FRAP1 (mTOR) is a serine⁄threonine kinase that plays a critical role in cellular growth and proliferation. Perturbations in the mTOR⁄PI3-kinase⁄AKT pathway are associated with numerous forms of cancer. FRAP1 is also the target of rapamycin and its analogues, which are currently used as immunosuppressants and cancer therapeutics.
Specifications

Specifications

NP_004949.1
Protein Assays and Analysis, Kinase Assay
163.9kDa
100μg
For research use only. Not for use in diagnostic procedures
FRAP1 (mTOR)
FRAP1 (mTOR) was pre-diluted in enzyme dilution buffer and assayed in 50mM HEPES (pH 7.5), 10mM MnCl2, 1mM EGTA, 2.5mM DTT, 0.01% Polysorbate 20, 100μM ATP, 500μg/mL PHAS-1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C
FRAP1 (mTOR) Recombinant Human Protein
Human
Baculovirus
80% by SDS-PAGE
0.45mg/mL
Frozen
FRAP1 (mTOR) Recombinant Human Protein
–80°C
1 Each
FRAP1 (mTOR) Recombinant Human Protein
PIKK kinase family
34nmoles of phosphate transferred to PHAS-1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 5μg/mL
GST Tag
Frozen
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.