Learn More
Brady™ FreezerBondz™ Ultra-thin Cryogenic Polyester Labels
Label curved frozen lab surfaces including tubes, vials and cryocanes with the ultra-thin B-492 polyester label material. Print almost anywhere with a durable Brady™ handheld label maker.
Brand:  Brady™ 170779
 View more versions of this product
Product Code. 17990730
  • £86.15 / Pack of 220
Estimated Shipment: 30-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ideal for laboratory use on curved surfaces including test tubes, vials and straw pipettes
  • Withstands chemicals, abrasions and extreme temperatures, including storage in liquid nitrogen
  • Made with a permanent acrylic adhesive for tight bonding onto smooth curved surfaces such as glass and polypropylene
Specifications
Specifications
Not Autoclavable
M4-130-492
White
Polyester
BMP41, BMP51, M511
-320°F to +230°F
0.375 x 0.825 in.
0.825 in.
0.5 in.
RoHS Compliant to 2011/65/EU (Pass 2015/863)
Permanent Acrylic
Die-cut
General Identification, Laboratory Labeling
220 Labels/Cartridge
0.0032 in.
0.375 in.
2.1 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Brady™ FreezerBondz™ Ultra-thin Cryogenic Polyester Labels > Dimensions (LxW): 0.375 x 0.825 in.; For Use With (Equipment): BMP41, BMP51, M511; Format: Die-cut