Learn More
Brady™ FreezerBondz™ Ultra-thin Cryogenic Polyester Laboratory Vial Top Labels for M6/M7 Printers
Durable Freezerbondz™ polyester markers are designed for laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws and slides.
Brand:  Brady™ 174114
67.40 GBP valid until 2024-12-20
Use promo code "21731" to get your promotional price.
 View more versions of this product
Product Code. 17828374
  • £79.80 / Pack of 500
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Can be applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen
  • Good print smudge resistance, solvent resistance and high/low-temperature performance range of -196°C to 100°C (-320°F to 212°F)
  • Made of B-492 polyester material
Specifications
Specifications
Not Autoclavable
Permanent Acrylic
White
Die-cut
Single-side Printable
Labels
General Identification, Laboratory Labeling
No
Rectangle with Circle
-196°C to +110°C
0.081 mm
0.375 in.
25.4 mm
0.375 in.
ASTM D 1000
None
Matte
Polyester
Thermal Transfer
Non-sterile
M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter)
500 Labels
-320°F to +230°F
0.0032 in.
0.375 x 1 in.
1 in.
9.53 mm
9.53 mm