Learn More
Labconco FreeZone 12L Console Freeze Dryer
FreeZone Console Freeze Dryers are designed for lyophilizing moderate to large sample loads or numerous small container batches. The console features a front viewing window, interior space for a vacuum pump, and accommodation for a large tray dryer.
£24695.00 - £59380.00
Specifications
Type | Console Freeze Dryer |
---|---|
Capacity (Metric) | 12 L |
Plug Type | All except North America |
Voltage | 230 V |
Product Code | Brand | Material | Temperature Range (Metric) | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Temperature Range (Metric) | Includes | Price | Quantity & Availability | |||
17921040
|
Labconco
701221090 |
Stainless Steel Coil and Collector Chamber | -50°C | Stoppering Tray Dryer |
£52465.00
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17971040
|
Labconco
701221190 |
Stainless Steel Coil and Collector Chamber | -50°C | Purge Valve |
£53885.00
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17981040
|
Labconco
701222090 |
PTFE Coil and Collector Chamber | -50°C | Stoppering Tray Dryer |
£53965.00
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17951050
|
Labconco
701222190 |
PTFE Coil and Collector Chamber | -50°C | Purge Valve, Stoppering Tray Dryer |
£55375.00
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17900180
|
Labconco
711211090 |
Stainless Steel Coil and Collector Chamber | -84°C |
£24695.00
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17950150
|
Labconco
711211190 |
Stainless Steel Coil and Collector Chamber | -84°C | Purge Valve |
£26110.00
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17990170
|
Labconco
711211290 |
Stainless Steel Coil and Collector Chamber | -84°C | Purge Valve, Mini Chamber |
£28535.00
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17910160
|
Labconco
711212090 |
PTFE Coil and Collector Chamber | -84°C |
£26200.00
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17980170
|
Labconco
711212190 |
PTFE Coil and Collector Chamber | -84°C | Purge Valve |
£27615.00
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17910140
|
Labconco
711212290 |
PTFE Coil and Collector Chamber | -84°C | Purge Valve, Mini Chamber |
£30040.00
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17971050
|
Labconco
711221090 |
Stainless Steel Coil and Collector Chamber | -84°C | Stoppering Tray Dryer |
£56455.00
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17901040
|
Labconco
711221190 |
Stainless Steel Coil and Collector Chamber | -84°C | Purge Valve, Stoppering Tray Dryer |
£57870.00
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17961040
|
Labconco
711222090 |
PTFE Coil and Collector Chamber | -84°C | Stoppering Tray Dryer |
£57960.00
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17981050
|
Labconco
711222190 |
PTFE Coil and Collector Chamber | -84°C | Purge Valve, Stoppering Tray Dryer |
£59380.00
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Coil holds 12 liters of ice before defrosting
- The hp HCFC/CFC-free refrigeration system cools the collector to -50° C (-58° F), suitable for aqueous samples.
- Includes a stoppering tray dryer with heated and cooled shelves, vacuum release valve control, stoppering knob and more.
- Lyo-Works™ OS provides a real time display of collector temperature, vacuum level and optional End-Zone™ end point detection system
- Large, intuitive 5 in. full color, capacitive touchscreen display for easy operation.
- Stay Connected
- View Real-Time Sample Data
- Get Consistent, High Quality Results
- Make Challenging Samples Easy
Specifications
Console Freeze Dryer | |
12 L | |
All except North America | |
230 V |