Learn More
Thermo Scientific™ Fresco™ 21 Microcentrifuge
GREENER_CHOICE
High-end research applications requiring microcentrifuges with refrigeration and performance up to 21,000xg.
Brand:  Thermo Scientific™ 75002555
4600.00 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.
 View more versions of this product
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Product Code. 15586334
  • £5750.00 / Each
Explore more special offers
Description
Description
Save time with the fast sample processing of Thermo Scientific Freso 21 microcentrifuges, providing productivity, versatility, safety and convenience in a compact, easy-to-use lab instrument. Lightweight microcentrifuge rotors are designed to provide acceleration up to 21,100xg in as little as 12 seconds. Refrigeration offers cooling from room temperature to set temperature in just 9 minutes.
Support your micro-volume separation needs with:
  • Exceptional acceleration of up to 21,000xg in as little as 12 seconds.
  • Refrigerated versions offer cooling from room temperature to set temperature in as little as 9 minutes.
  • Choose from 7 optional microcentrifuge rotors to find the ideal fit for your application:
    • Outstanding corrosion resistance with lightweight engineered polymer rotors.
    • Standard rotor runs 24 microcentrifuge tubes in a single row, from 1.5 to 2.0 mL tubes to mini-preps and spin columns.
    • Ergonomic one-click rotor opening and closing, as well as transparent design for added safety and convenience with Thermo Scientific ClickSeal biocontainment lids.
    • Eliminate the need for adapters with unique dual row rotor.
  • Simplify operation with intuitive controls, easy-to-read displays and fast one-click centrifuge lid closure.
  • Sleek, lightweight, space-saving design.
  • Durable and easy to clean and maintain.
  • Quiet performance promotes a safe, stress-free environment.
  • Conforms to the latest clinical and safety standards, including 98/79/EC In Vitro Diagnostic Directive.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
24 x 1.5/2 mL
21,100 x g
-9°C to +40°C
Benchtop Centrifuge
Fresco 21 Centrifuge, Refrigerated
Yes
IEC 61010-1, IEC 61010-2-020 (refer to manual for more details)
62.0lb. (28.0 kg)
50/60 Hz
Dual Row 18 x 2/0.5 mL Rotor with Screw-On Lid
14,800 rpm
50 dB(A)
CSA, CE, IVD
Dual Row 18 x 2.0/0.5mL rotor with screw-on lid
230 V 50/60 Hz
1 to 99 min. (1 min. increments); HOLD
12.9 x 11.6 x 17.5 in. (33.0 x 29.5 x 44.5 cm)
Thermo Scientific
230 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products