Learn More
Fritsch™ Silicon Nitride Grinding Balls
Brand:  Fritsch™ 55.0300.31
Encompass
 View more versions of this product
Product Code. 13161674
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Specifications
Grinding Ball
3.25 g/cm3
For abrasive samples, metal-free grinding
91.1HRA
Abrasive, metal-free grinding
Al2O3Y2O3: 12.5share%, Si3N4: 86.5share%, Other: 1.00share%
30 mm
For planetary mono mill Pulverisette 6 classic line
Silicon Nitride