missing translation for 'onlineSavingsMsg'
Learn More

Carbolite™ Furnace with Type R Temperature Sensor

Brand:  Carbolite™ RHF1403-230SN+&02-301


Code : 04

Additional Details : Weight : 100.00000kg

 View more versions of this product

Product Code. 10551305

  • £6315.00 / Each
Estimated Shipment: 11-09-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Compliance to BSEN 61010
3 L
Furnace
200 x 120 x 120 mm
120 mm
120 mm
200 mm
230 V
50/60 Hz
301 PID Controller
1400°C
For use in quality control, heat treatment and research of ceramic materials.
435 mm
655 mm
610 mm
230 V 50/60 Hz 4500 w
4500 w
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
View accessory products
Special Offers

Special Offers