missing translation for 'onlineSavingsMsg'
Learn More

Carbolite™ Furnace with Type R Temperature Sensor

Brand:  Carbolite™ HTF1705-230SN+TMP-3216P1BASE


Code : 04

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 12089872

  • £14020.00 / Each
Estimated Shipment: 10-09-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Compliance to BSEN 61010
5 L
Furnace
225 x 150 x 158 mm
150 mm
158 mm
225 mm
230 V
50/60 Hz
3216P1 Controller
1700°C
For use in quality control, heat treatment and research of ceramic materials.
830 mm
565 mm
650 mm
230 V 50/60 Hz 4500 w
4500 w
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers