missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
CAS | 54-31-9 |
---|---|
Molecular Formula | C12H11ClN2O5S |
Molecular Weight (g/mol) | 330.74 |
InChI Key | ZZUFCTLCJUWOSV-UHFFFAOYSA-N |
Synonym | furosemide, frusemide, lasix, furosemid, furanthril, errolon, fusid, aisemide, beronald, desdemin |
PubChem CID | 3440 |
ChEBI | CHEBI:47426 |
IUPAC Name | 4-chloro-2-(furan-2-ylmethylamino)-5-sulfamoylbenzoic acid |
SMILES | C1=COC(=C1)CNC2=CC(=C(C=C2C(=O)O)S(=O)(=O)N)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
13591610
|
Thermo Scientific Chemicals
448970010 |
1 g | Glass bottle |
£37.08
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13591640
|
Thermo Scientific Chemicals
448970050 |
5 g | Glass bottle |
£121.61
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
54-31-9 | |
330.74 | |
furosemide, frusemide, lasix, furosemid, furanthril, errolon, fusid, aisemide, beronald, desdemin | |
CHEBI:47426 | |
C1=COC(=C1)CNC2=CC(=C(C=C2C(=O)O)S(=O)(=O)N)Cl |
C12H11ClN2O5S | |
ZZUFCTLCJUWOSV-UHFFFAOYSA-N | |
3440 | |
4-chloro-2-(furan-2-ylmethylamino)-5-sulfamoylbenzoic acid |
Specifications
54-31-9 | |
97% min. (HPLC) | |
15,4338 | |
Solubility in water: slightly soluble | |
C1=COC(=C1)CNC2=CC(=C(C=C2C(=O)O)S(=O)(=O)N)Cl | |
330.74 | |
CHEBI:47426 | |
97+% |
218.0°C to 220.0°C | |
C12H11ClN2O5S | |
furosemide, frusemide, lasix, furosemid, furanthril, errolon, fusid, aisemide, beronald, desdemin | |
ZZUFCTLCJUWOSV-UHFFFAOYSA-N | |
4-chloro-2-(furan-2-ylmethylamino)-5-sulfamoylbenzoic acid | |
3440 | |
330.74 | |
Furosemide |
Safety and Handling
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-203-6
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Furosemide
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360-May damage fertility or the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title