missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Furosemide, 97+% Product Code.: 13591610

Thermo Scientific Chemicals Furosemide, 97+%

Product Code. 13591610
1 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 13591610

Brand: Thermo Scientific Chemicals 448970010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-31-9
Molecular Formula C12H11ClN2O5S
Molecular Weight (g/mol) 330.74
InChI Key ZZUFCTLCJUWOSV-UHFFFAOYSA-N
Synonym furosemide, frusemide, lasix, furosemid, furanthril, errolon, fusid, aisemide, beronald, desdemin
PubChem CID 3440
ChEBI CHEBI:47426
IUPAC Name 4-chloro-2-(furan-2-ylmethylamino)-5-sulfamoylbenzoic acid
SMILES C1=COC(=C1)CNC2=CC(=C(C=C2C(=O)O)S(=O)(=O)N)Cl

Specifications

Melting Point 218.0°C to 220.0°C
Assay Percent Range 97% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Merck Index 15,4338
Solubility Information Solubility in water: slightly soluble
Formula Weight 330.74
Percent Purity 97+%
Chemical Name or Material Furosemide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0420078 Certificate TypeCertificate of Analysis Date30/12/2022 Catalog Number448970050,448970010
Lot NumberA0448148 Certificate TypeCertificate of Analysis Date30/11/2022 Catalog Number448970050,448970010
compliance-icons
Product Identifier
  • Furosemide
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Furosemide, 97+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.