Learn More
A&D Weighing FX/FZ Precision Balances
A&D Weighing FZ/FX Series Precision Balances combine all of the essential weighing functions in a compact, light-weight package.
£753.00 - £1030.00
Specifications
Certifications/Compliance | GLP/GMP/GCP/ISO |
---|---|
Display Type | VFD |
Material | Stainless Steel |
Stabilization Time | 1 second |
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
16601072
![]() |
A&D Weighing
FX-120i |
122 g |
£777.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16601082
![]() |
A&D Weighing
FX-5000i |
5 kg |
£1005.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16611082
![]() |
A&D Weighing
FX-500i |
520 g |
£1030.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16621072
![]() |
A&D Weighing
FX-2000i |
2200 g |
£840.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16641072
![]() |
A&D Weighing
FX-200i |
220 g |
£757.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16661072
![]() |
A&D Weighing
FX-3000i |
3200 g |
£891.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16681062
![]() |
A&D Weighing
FX-1200i |
1220 g |
£753.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16681072
![]() |
A&D Weighing
FX-300i |
320 g |
£896.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Built-in internal calibration at the touch of a button
- Shock absorber function
- Auto power ON/OFF function
- B5 size footprint for use in narrow spaces
- VFD Display for easy to read weighing results
- SCF – Statistical Calculation Function
- Standard RS-232C
- Fast compact SHS sensor, response time only 1 second
- Quick USB interface – no driver installation required (optional)
- LAN – Ethernet interface with WinCT-Plus software (optional)
- Animal weighing pan (optional)
- Large breeze break (optional)
- Small breeze break included on 1 mg models
- Built-in rechargeable NiMH battery pack (optional)
Specifications
GLP/GMP/GCP/ISO | |
VFD | |
Stainless Steel | |
1 second |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
A&D Weighing FX/FZ Precision Balances