Learn More
A&D Weighing FX/FZ Precision Balances IP 65
A&D Weighing FZ/FX Series Precision Balances combine all of the essential weighing functions in a compact, light-weight package.
Brand:  A&D Weighing FX-200i-WP
 View more versions of this product
Product Code. 16651072
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • IP65 water / dust proof
  • Built-in internal calibration at the touch of a button
  • Shock absorber function
  • Auto power ON/OFF function
  • B5 size footprint for use in narrow spaces
  • VFD Display for easy to read weighing results
  • SCF – Statistical Calculation Function
  • Standard RS-232C
  • Fast compact SHS sensor, response time only 1 second
  • Quick USB interface – no driver installation required (optional)
  • LAN – Ethernet interface with WinCT-Plus software (optional)
  • Animal weighing pan (optional)
  • Large breeze break (optional)
  • Small breeze break included on 1 mg models
  • Built-in rechargeable NiMH battery pack (optional)
Specifications
Specifications
GLP/GMP/GCP/ISO/IP65
220 g
0.001 g
FX-200i-WP
11 VA to the AC adaptor (12 V, 1 A)
RS-232 C, Quick USB (option), LAN (option)
VFD
Stainless Steel
1 second
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
A&D Weighing FX/FZ Precision Balances IP 65 > 220 g x 1 mg