Learn More
A&D Weighing FX/FZ Precision Balances IP 65
GREENER_CHOICE
A&D Weighing FZ/FX Series Precision Balances combine all of the essential weighing functions in a compact, light-weight package.
£995.00 - £1415.00
Specifications
Certifications/Compliance GLP/GMP/GCP/ISO/IP65
Connection Type RS-232 C, Quick USB (option), LAN (option)
Display Type VFD
Material Stainless Steel
Stabilization Time 1 second
View More Specs
Products 5
Product Code Brand Capacity Price Quantity & Availability  
Product Code Brand Capacity Price Quantity & Availability  
16631072
GREENER_CHOICE
View Documents
A&D Weighing
FX-2000i-WP
2200 g
£1215.00
Each
16651072
View Documents
A&D Weighing
FX-200i-WP
220 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16671072
GREENER_CHOICE
View Documents Promotion Details
A&D Weighing
FX-3000i-WP
3200 g
£1415.00
Each
16691062
GREENER_CHOICE
View Documents
A&D Weighing
FX-1200i-WP
1220 g
£1155.00
Each
16691072
GREENER_CHOICE
View Documents
A&D Weighing
FX-300i-WP
320 g
£1280.00
Each
Description
Description
  • IP65 water / dust proof
  • Built-in internal calibration at the touch of a button
  • Shock absorber function
  • Auto power ON/OFF function
  • B5 size footprint for use in narrow spaces
  • VFD Display for easy to read weighing results
  • SCF – Statistical Calculation Function
  • Standard RS-232C
  • Fast compact SHS sensor, response time only 1 second
  • Quick USB interface – no driver installation required (optional)
  • LAN – Ethernet interface with WinCT-Plus software (optional)
  • Animal weighing pan (optional)
  • Large breeze break (optional)
  • Small breeze break included on 1 mg models
  • Built-in rechargeable NiMH battery pack (optional)
Specifications
Specifications
GLP/GMP/GCP/ISO/IP65
VFD
1 second
RS-232 C, Quick USB (option), LAN (option)
Stainless Steel
11 VA to the AC adaptor (12 V, 1 A)
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
A&D Weighing FX/FZ Precision Balances IP 65