Learn More
GeneBLAzer™ Gα15-NFAT CHO-K1 DA Assay Kit
GeneBLAzer™ Gα15-NFAT CHO-K1 DA Assay Kit contains the promiscuous G protein.
Brand: GeneBLAzer™ K1453
Code : NEW
Additional Details : Weight : 0.05000kg
Description
- Gene Symbol: Gα15
- Includes: GeneBLAzer™ Gα15-NFAT CHO-K1 DA cells, LiveBLAzer™-FRET B/G Loading Kit, 70μg, Solution D, GeneBLAzer™ Gα15-NFAT CHO-K1 Assay Protocol, LiveBLAzer™-FRET B/G Loading Protocol, Certificate of Analysis
- The GeneBLAzer™ Gα15-NFAT CHO-K1 DA (Division Arrested) cell line contains the promiscuous G protein, Gα15, stably integrated into the CellSensor™ NFAT-bla CHO-K1 (Cat #K1078) cell line. NFAT-bla CHO-K1 cells contain a beta-lactamase reporter gene under control of an NFAT response element stably integrated into CHO-K1 cells. DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin C, and have no apparent toxicity or change in cellular signal transduction.
- This product can serve as a negative control in screening assays performed with specific G-protein coupled receptor (GPCR) DA cells that were made from this same background. These cells can also be used in other assays where a sensitive readout to intracellular changes in calcium is needed.
Specifications
Liquid Nitrogen | |
GPCRs | |
Division-Arrested Cells Assay Kit |
Fluorescent | |
1 Plate |