missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) 2,4-pentanedionate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

£58.29 - £206.03

Chemical Identifiers

CAS 64438-54-6
Molecular Formula C15H21GdO6
Molecular Weight (g/mol) 454.58
MDL Number MFCD00013493
InChI Key PJCXQIZIMGZZIT-UHFFFAOYSA-N
Synonym gadolinium iii 2,4-pentanedionate hydrate, reacton, r reo
PubChem CID 131675701
IUPAC Name gadolinium(3+);(Z)-4-oxopent-2-en-2-olate;hydrate
SMILES [Gd+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11396648
SDS View Documents
Thermo Scientific Alfa Aesar
013204.06
5 g
£58.29
5g
Please to purchase this item. Need a web account? Register with us today!
11386648
SDS View Documents
Thermo Scientific Alfa Aesar
013204.14
25 g
£206.03
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used as a laboratory chemicals & for manufacture of chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gadolinium(III) 2,4-pentanedionate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.