missing translation for 'onlineSavingsMsg'
Learn More
Gadolinium(III) nitrate hexahydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals Product Code.: 10687504

Gadolinium(III) nitrate hexahydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals

Product Code. 10687504
100 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10687504

Brand: Thermo Scientific Acros 316761000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19598-90-4
Molecular Formula GdN3O9·6H2O
Molecular Weight (g/mol) 451.4
MDL Number MFCD00149728
InChI Key XWFVFZQEDMDSET-UHFFFAOYSA-N
Synonym gadolinium iii nitrate hexahydrate, gadolinium nitrate hexahydrate, gadolinium iii nitrate, hexahydrate 1:3:6, nitric acid, gadolinium 3+ salt, hexahydrate, gadoliniumnitratehexahydrate, gd.3no3.6h2o, gadolinium 3+ trinitrate hexahydrate, gadolinium nitrate, hexahydrate,, nitric acid,gadolinium 3+ salt, hexahydrate 8ci,9ci, gadolinium iii nitrate hexahydrate trace metals basisShow MoreShow Less
PubChem CID 209258
IUPAC Name gadolinium(3+);trinitrate;hexahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Gd+3]

Specifications

Melting Point 91°C
Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 100 g
Assay Percent Range 99.90%
Linear Formula Gd(NO3)3·6H2O
Solubility Information Solubility in water: soluble
Formula Weight 451.4
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Gadolinium(III) nitrate hexahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467871 Certificate TypeCertificate of Analysis Date29/11/2024 Catalog Number316761000,316760250
Lot NumberA0464718 Certificate TypeCertificate of Analysis Date19/07/2024 Catalog Number316761000,316760250
Lot NumberA0461748 Certificate TypeCertificate of Analysis Date10/04/2024 Catalog Number316761000,316760250
Lot NumberA0456669 Certificate TypeCertificate of Analysis Date03/10/2023 Catalog Number316761000,316760250
Lot NumberA0427012 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number316761000,316760250
compliance-icons compliance-icons
Product Identifier
  • Gadolinium(III) nitrate hexahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
Supplemental information
  • MIXTURE LIST-Contains : Gadolinium(III) nitrate hexahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gadolinium(III) nitrate hexahydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.