Learn More
Gadolinium(III) oxide, nanopowder, 99.99+% (REO), Thermo Scientific Chemicals
Description
Gadolinium(III) oxide is used as a raw material for various fluorescent compounds, absorption materials in atomic reactions, nuclear fuels, magnetic bubble material and screen-sensitivity increasing material. It is also used in glass and electronic industries. It also acts as a neutron shield, a catalyst, a dielectric ceramic and is used for filament coatings, special glasses, lasers, masers and telecommunication. It also serves as a laboratory reagent. It plays an important role as a potential contrast agent for magnetic resonance imaging (MRI).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 2330°C |
Physical Form | Powder |
Quantity | 100 g |
Merck Index | 14,4326 |
Solubility Information | Soluble in acid. Insoluble in water. |
Formula Weight | 362.5 |
Percent Purity | ≥99.99% |
Density | 7.407 g/mL |
Assay | (REO) |
Chemical Name or Material | Gadolinium (III) oxide |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P17H005 | Certificate of Analysis | 31/01/2025 | 044020.22,044020.14,044020.36 |
M20I009 | Certificate of Analysis | 12/04/2022 | 044020.22,044020.14,044020.36 |
C01Y011 | Certificate of Analysis | 22/11/2021 | 044020.22,044020.14,044020.36 |
B21T042 | Certificate of Analysis | 22/11/2021 | 044020.22,044020.14,044020.36 |
C24T038 | Certificate of Analysis | 22/11/2021 | 044020.22,044020.14,044020.36 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.