missing translation for 'onlineSavingsMsg'
Learn More
Gadolinium(III) oxide, REacton™, 99.999% (REO), Thermo Scientific Chemicals Product Code.: 11321019

Gadolinium(III) oxide, REacton™, 99.999% (REO), Thermo Scientific Chemicals

Product Code. 11321019
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11321019

Brand: Thermo Scientific Alfa Aesar 011289.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gadolinium(III) oxide is used as a raw material for various fluorescent compounds, absorption material in atomic reactions, nuclear fuels, magnetic bubble material and screen-sensitivity increasing material. It is also used in glass and electronic industries. It also acts as a neutron shield, a catalyst, a dielectric ceramic and is used for filament coatings, special glasses, lasers, masers and telecommunication. It also serves as a laboratory reagent. It plays an important role as a potential contrast agent for magnetic resonance imaging (MRI).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12064-62-9
Molecular Formula Gd2O3
Molecular Weight (g/mol) 362.50
MDL Number MFCD00011026
InChI Key CMIHHWBVHJVIGI-UHFFFAOYSA-N
Synonym gadolinia, gadolinium oxide, gadolinium iii oxide, gadolinium trioxide, digadolinium trioxide, gadolinium sesquioxide, gadolinium 3+ oxide, unii-5480d0nhlj, gadolinium 3+ ; oxygen 2-, digadolinium 3+ ion trioxidandiideShow MoreShow Less
PubChem CID 159427
SMILES [O--].[O--].[O--].[Gd+3].[Gd+3]

Specifications

Melting Point 2330°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.999% (REO)
Solubility Information Soluble in acid. Insoluble in water.
IUPAC Name digadolinium(3+) trioxidandiide
Formula Weight 362.5
Appearance White powder or granules
Density 7.407 g/cm3
Chemical Name or Material Gadolinium(III) oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250999 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog Number011289.22,011289.14,011289.36
Lot NumberD09S014 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number011289.22,011289.14,011289.36
Lot NumberG23W055 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number011289.22,011289.14,011289.36
Lot NumberL11R010 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number011289.22,011289.14,011289.36
Lot NumberE11S024 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number011289.22,011289.14,011289.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gadolinium(III) oxide, REacton™, 99.999% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.