missing translation for 'onlineSavingsMsg'
Learn More
Gadolinium powder, -40 mesh, 99% (REO), Thermo Scientific Chemicals Product Code.: 15461027

Gadolinium powder, -40 mesh, 99% (REO), Thermo Scientific Chemicals

Product Code. 15461027
2 g
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy

Product Code. 15461027

Brand: Thermo Scientific Alfa Aesar 012896.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-54-2
Molecular Formula Gd
Molecular Weight (g/mol) 157.25
MDL Number MFCD00011022
InChI Key UIWYJDYFSGRHKR-UHFFFAOYSA-N
Synonym atom, gadolinio, unii-au0v1lm3jt, trihydride, au0v1lm3jt, foil, 3n, gadolinium, elemental, ingot, gadolinium, chips, compounds
PubChem CID 23982
ChEBI CHEBI:33375
IUPAC Name gadolinium
SMILES [Gd]

Specifications

Color Silver
UN Number UN3178
Quantity 2 g
Formula Weight 157.25
Percent Purity 99%
Mesh Size −40 Mesh
Form Powder
Assay (REO)
Chemical Name or Material Gadolinium powder

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY28I010 Certificate TypeCertificate of Analysis Date17/01/2023 Catalog Number012895.18,012895.09,012896.04
Lot NumberX06I026 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog Number012895.18,012895.09,012896.04
Lot NumberP15I029 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number012895.18,012895.09,012896.04
Lot NumberP14I028 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number012895.18,012895.09,012896.04
Lot NumberP15I024 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number012895.18,012895.09,012896.04

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gadolinium powder, -40 mesh, 99% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.