missing translation for 'onlineSavingsMsg'
Learn More
Gallium(III) nitrate hydrate, 99.9998%, (trace metal basis), Thermo Scientific Chemicals Product Code.: 10319143

Gallium(III) nitrate hydrate, 99.9998%, (trace metal basis), Thermo Scientific Chemicals

Product Code. 10319143
25 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10319143

Brand: Thermo Scientific Acros 212440250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gallium(III) nitrate hydrate, 99.9998%, (trace metal basis), ACROS Organics, Quantity: 100g, Packaging: Glass bottle, CAS: 69365-72-6, White, Grade: Trace Metal Basis, Melting Point: 110.0 deg.C, Molecular Formula: GaN3O9.xH2O, Molecular Weight: 255.74, Percent Purity: 99.9998%

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69365-72-6
Molecular Formula GaN3O9
Molecular Weight (g/mol) 255.74
MDL Number MFCD00149723
InChI Key CHPZKNULDCNCBW-UHFFFAOYSA-N
Synonym gallium iii nitrate hydrate, nitric acid, gallium salt, hydrate, ga.3no3.h2o, gallium nitrate nanoparticles, gallium-iii-nitrate hydrate, gallium iii nitrate hydrate, puratronic, gallium iii nitrate hydrate, puratronic metals basis, gallium iii nitrate hydrate, crystalline trace metals basis, gallium iii nitrate hydrate, crystals and lumps trace metals basisShow MoreShow Less
PubChem CID 11550823
SMILES [Ga+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 110.0°C
Color White
Physical Form Crystalline Powder, Crystals and/or Chunks
Quantity 25 g
Assay Percent Range 99.9998%
Linear Formula Ga(NO3)3·xH2O
Merck Index 14, 4350
Solubility Information Solubility in water: soluble
IUPAC Name gallium(3+) trinitrate
Formula Weight 255.74
Percent Purity 99.9998%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Gallium(III) nitrate hydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0403447 Certificate TypeCertificate of Analysis Date07/12/2023 Catalog Number212440050,212440010,212441000,212440250
Lot NumberA0456253 Certificate TypeCertificate of Analysis Date12/09/2023 Catalog Number212440050,212440010,212441000,212440250
Lot NumberA0449096 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number212440050,212440010,212441000,212440250
Lot NumberA0398459 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number212440050,212440010,212441000,212440250
Lot NumberA0436346 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number212440050,212440010,212441000,212440250
compliance-icons compliance-icons
Product Identifier
  • Gallium(III) nitrate hydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gallium(III) nitrate hydrate, 99.9998%, (trace metal basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.