missing translation for 'onlineSavingsMsg'
Learn More
Gallium(III) oxide, 99.99+%, (trace metal basis), Thermo Scientific Chemicals Product Code.: 10780965

Gallium(III) oxide, 99.99+%, (trace metal basis), Thermo Scientific Chemicals

Product Code. 10780965
50 g, Plastic bottle
Click to view available options
Quantity:
5 g
50 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10780965

Brand: Thermo Scientific 201100500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12024-21-4
Molecular Formula Ga2O3
Molecular Weight (g/mol) 187.44
MDL Number MFCD00011020
InChI Key AJNVQOSZGJRYEI-UHFFFAOYSA-N
IUPAC Name digallium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Ga+3].[Ga+3]

Specifications

Melting Point 1740°C
Color White
Physical Form Fine Crystalline Powder
Quantity 50 g
Assay Percent Range 99.99+%
Linear Formula Ga2O3
Fieser 04,241
Merck Index 15, 4382
Solubility Information Solubility in water: insoluble in water. Other solubilities: slightly soluble in acids,alkalies
Formula Weight 187.44
Percent Purity ≥99.99%
Grade Trace Metal Basis
Packaging Plastic bottle
Specific Gravity 5.88
Density 5.8800g/mL
Chemical Name or Material Gallium(III) oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462796 Certificate TypeCertificate of Analysis Date22/05/2024 Catalog Number201100050,201100500
Lot NumberA0456892 Certificate TypeCertificate of Analysis Date12/10/2023 Catalog Number201100050,201100500
Lot NumberA0454374 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number201100050,201100500
Lot NumberA0403517 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number201100050,201100500
Lot NumberA0399765 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number201100050,201100500
Product Identifier
  • Gallium(III) oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gallium(III) oxide, 99.99+%, (trace metal basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.