missing translation for 'onlineSavingsMsg'
Learn More

WTW™ Galvanic Dissolved Oxygen Sensors (universal)

Get accurate testing results for water and waterous solutions with these membrane-covered galvanic dissolved oxygen sensors that are low maintenance and no polarizarion time required.

£694.00 - £1229.20

Specifications

For Use With (Equipment) Conductivity and multiparameter meters
View More Specs

Products 4
Product Code Brand Length (Metric) Cable Includes Price Quantity & Availability  
Product Code Brand Length (Metric) Cable Includes Price Quantity & Availability  
10503833
View accessory products View Documents
WTW™
201570
3 m 3m L Cable Cleaning solution, abrasive foil, 1 exchange membrane head and electrolyte
£694.00
Each
Estimated Shipment: 26-03-2025
to see stock.
Add to basket
 
10776701
View accessory products View Documents
WTW™
201548
15 m 15m Length Cable Cleaning solution, abrasive foil, 3 exchange membrane heads and electrolyte This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10470632
View accessory products View Documents
WTW™
201545
3 m 3m L Cable Cleaning solution, abrasive foil, 3 exchange membrane heads and electrolyte
£1030.00
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
 
10491592
View accessory products View Documents
WTW™
201546
6 m 6m Length Cable Cleaning solution, abrasive foil, 3 exchange membrane heads and electrolyte
£1015.00
Each
Estimated Shipment: 26-03-2025
to see stock.
Add to basket
 
Description

Description

  • Dissolved oxygen sensor with integrated temperature compensation and membrane leak monitoring
  • Can be used in the lab as well as in the field
  • Features include a quick response time (t99<60 s) and low-maintenance operation
  • Fields of use include surface waters, measurements in wastewater treatment plants, BOD directions
  • Filled with electrolyte and achieves a long dwelling time up to 6 months
  • Calibration vessel is part of the delivery
  • For use with conductivity and multiparameter meters
  • Includes 1.5m L Cable Cleaning solution, abrasive foil, 3 exchange membrane heads and electrolyte
 
Specifications

Specifications

Conductivity and multiparameter meters
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.