Learn More
Keystone Cleanroom Products Gamma-Irradiated Fine Point Sharpie
Ready-to-use gamma irradiated pens and markers eliminate the risks associated with surface sanitization procedures.
Brand: Keystone Cleanroom Products S01FMARKBLUE
Additional Details : Weight : 11.22640kg
Description
- Gamma irradiated
- Lot controlled
- Ready to use
- Packaged in easy to open Tyvek™ peel pouches
- Minimize contamination risks
- Simplifies conformance to SOP's
- Eliminate the need for wiping down contaminated writing instruments
Specifications
Plastic | |
14 cm | |
COC, Certificate of Irradiation | |
Sterile |
Blue | |
Not Autoclavable | |
Yes |