missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14305-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the cytoskeletal BTB/kelch repeat family. The encoded protein plays a role in neurofilament architecture and is involved in mediating the ubiquitination and degradation of some proteins. Defects in this gene are a cause of giant axonal neuropathy.Specifications
GAN | |
Polyclonal | |
Unconjugated | |
GAN | |
GAN, GAN1, gigaxonin, Kelch like protein 16, KLHL16 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
209239, 307893, 8139 | |
-20°C | |
Liquid |
Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8CA72, Q9H2C0 | |
GAN | |
GAN Fusion Protein Ag5421 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |