missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | GAN |
---|---|
Concentration | 0.28 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16852884
|
Proteintech
14305-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16842884
|
Proteintech
14305-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a member of the cytoskeletal BTB/kelch repeat family. The encoded protein plays a role in neurofilament architecture and is involved in mediating the ubiquitination and degradation of some proteins. Defects in this gene are a cause of giant axonal neuropathy.Specifications
GAN | |
Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
Q8CA72, Q9H2C0 | |
209239, 307893, 8139 | |
GAN Fusion Protein Ag5421 | |
Primary | |
-20°C | |
GAN |
0.28 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
GAN, GAN1, gigaxonin, Kelch like protein 16, KLHL16 | |
GAN | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |